missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromobenzylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
104.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a intermediate for pharmaceutical.
Solubility
Reacts with water.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-vent
It is employed as a intermediate for pharmaceutical.
Solubility
Reacts with water.
Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-vent
Specifications
CAS | 307496-31-7 |
Density | 1.027 |
Molecular Formula | C7H6Br2Zn |
MDL Number | MFCD01311434 |
Quantity | 50 mL |
UN Number | UN3399 |
Sensitivity | Air sensitive |
Synonym | 3-bromobenzylzinc bromide, bromo 3-bromophenyl methyl zinc, 3-bromobenzylzinc bromide, 0.50 m in thf, 3-bromobenzylzinc bromide 0.5 m in tetrahydrofuran, 3-bromobenzylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles |
Solubility Information | Reacts with water. |
InChI Key | FBSUMSXZNCNBLE-UHFFFAOYSA-M |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'