missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzylsulfonyl chloride, 97%, Thermo Scientific™

Catalog Number p-3522487
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
58236-74-1
C7H6BrClO2S
269.537
MFCD06798043
VCGKEAIINMDLMC-UHFFFAOYSA-N
3-bromophenyl methanesulfonyl chloride, 3-bromobenzylsulfonyl chloride, 3-bromophenyl methanesulphonyl chloride, benzenemethanesulfonylchloride, 3-bromo, benzenemethanesulfonyl chloride, 3-bromo, 3-bromobenzylsulphonyl chloride, 3-bromophenyl methylsulphonyl chloride
2761451
(3-bromophenyl)methanesulfonyl chloride
C1=CC(=CC(=C1)Br)CS(=O)(=O)Cl
This item is not returnable. View return policy
58236-74-1
C7H6BrClO2S
269.537
MFCD06798043
VCGKEAIINMDLMC-UHFFFAOYSA-N
3-bromophenyl methanesulfonyl chloride, 3-bromobenzylsulfonyl chloride, 3-bromophenyl methanesulphonyl chloride, benzenemethanesulfonylchloride, 3-bromo, benzenemethanesulfonyl chloride, 3-bromo, 3-bromobenzylsulphonyl chloride, 3-bromophenyl methylsulphonyl chloride
2761451
(3-bromophenyl)methanesulfonyl chloride
C1=CC(=CC(=C1)Br)CS(=O)(=O)Cl
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 269.55
Percent Purity 97%
Chemical Name or Material 3-Bromobenzylsulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'