missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzenesulfonyl chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7081504
29.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2905-24-0
C6H4BrClO2S
255.51
MFCD00052313
PJGOLCXVWIYXRQ-UHFFFAOYSA-N
3-bromobenzene-1-sulfonyl chloride, 3-bromobenzenesulphonyl chloride, m-bromobenzenesulfonyl chloride, 3-bromobenzenesulfonylchloride, m-bromobenzenesulphonyl chloride, 3-bromo-benzenesulfonyl chloride, 3-bromophenyl chlorosulfone, 3-bromophenylsulfonyl chloride, buttpark 94\57-33, benzenesulfonyl chloride, 3-bromo
17943
3-bromobenzenesulfonyl chloride
ClS(=O)(=O)C1=CC=CC(Br)=C1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2905-24-0
C6H4BrClO2S
255.51
MFCD00052313
PJGOLCXVWIYXRQ-UHFFFAOYSA-N
3-bromobenzene-1-sulfonyl chloride, 3-bromobenzenesulphonyl chloride, m-bromobenzenesulfonyl chloride, 3-bromobenzenesulfonylchloride, m-bromobenzenesulphonyl chloride, 3-bromo-benzenesulfonyl chloride, 3-bromophenyl chlorosulfone, 3-bromophenylsulfonyl chloride, buttpark 94\57-33, benzenesulfonyl chloride, 3-bromo
17943
3-bromobenzenesulfonyl chloride
ClS(=O)(=O)C1=CC=CC(Br)=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 30°C to 33°C
Density 1.773
Boiling Point 90°C to 91°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.593
Quantity 1 g
UN Number UN3261
Beilstein 2691656
Sensitivity Moisture sensitive
Formula Weight 255.52
Percent Purity 97%
Chemical Name or Material 3-Bromobenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'