missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7021360
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
591-19-5
C6H6BrN
172.03
MFCD00007757
DHYHYLGCQVVLOQ-UHFFFAOYSA-N
m-bromoaniline, benzenamine, 3-bromo, 3-bromo aniline, m-aminobromobenzene, aniline, m-bromo, 3-bromobenzenamine, 3-bromophenylamine, bromoaniline 3-, 3-bromo-benzenamine, 3-bromo-phenylamine
11562
3-bromoaniline
NC1=CC=CC(Br)=C1
This item is not returnable. View return policy
591-19-5
C6H6BrN
172.03
MFCD00007757
DHYHYLGCQVVLOQ-UHFFFAOYSA-N
m-bromoaniline, benzenamine, 3-bromo, 3-bromo aniline, m-aminobromobenzene, aniline, m-bromo, 3-bromobenzenamine, 3-bromophenylamine, bromoaniline 3-, 3-bromo-benzenamine, 3-bromo-phenylamine
11562
3-bromoaniline
NC1=CC=CC(Br)=C1

3-Bromoaniline is used in the synthesis of amino substituted quinazoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromoaniline is used in the synthesis of amino substituted quinazoline.

Solubility
Soluble in water (0.97 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 16°C
Density 1.580
Boiling Point 251°C
Flash Point >110°C (230°F)
Assay Percent Range 98%
Refractive Index 1.625
Quantity 25 g
UN Number UN2810
Beilstein 742028
Solubility Information Soluble in water (0.97g/L at 20°C).
Formula Weight 172.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromoaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'