missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-phenyl-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64920.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15419256

  • £351.92 / 5g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-phenyl-5-(trifluoromethyl)benzenesulfonamide
380.18
MFCD27665324

Specifications

3-Bromo-N-phenyl-5-(trifluoromethyl)benzenesulfonamide
5g
380.18
Solid
97%
MFCD27665324
Videos
SDS