Alfa Aesar™ 3-Bromo-N-n-hexyl-5-(trifluoromethyl)benzenesulfonamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64356.MD

250MG 3-Bromo-N-n-hexyl-5-(trifluoromethyl)benzenesulfonamide, 97% 250mg

UNSPSC: 12352105

Code: NEW

Additional Details:
Additional Details: CAS Number: 951884-63-2 Weight: 0.00025kg


Quantity 250mg

Product Code. 15448866

Quantity Price
1 £ 18.66 / 250mg
Estimated Shipment date
from Supplier 06-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3-Bromo-N-n-hexyl-5-(trifluoromethyl)benzenesulfonamide
CAS 951884-63-2
Formula Weight 288.24
MDL Number MFCD09801008

Specification

Formula Weight 288.24
Chemical Name or Material 3-Bromo-N-n-hexyl-5-(trifluoromethyl)benzenesulfonamide
Physical Form Solid
Quantity 250mg
Assay 97%
MDL Number MFCD09801008
CAS 951884-63-2