missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-cyclohexyl-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64693.03

UNSPSC : 12352105

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419076

  • £70.28 / 1g


Estimated Shipment date
from Supplier 11-07-2017
Add to basket

Specifications Description & Specifications

Chemical Identifiers

3-Bromo-N-cyclohexyl-5-(trifluoromethyl)benzenesulfonamide
386.23
MFCD27665304

Specifications

3-Bromo-N-cyclohexyl-5-(trifluoromethyl)benzenesulfonamide
1g
386.23
Solid
97%
MFCD27665304
Documentation

Technical Documents