missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(4-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64890.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15439236

  • £79.71 / 1g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(4-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
394.21
MFCD27665321
3-Bromo-N-(p-tolyl)-5-(trifluoromethyl)benzenesulfonamide

Specifications

3-Bromo-N-(4-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
Solid
394.21
3-Bromo-N-(p-tolyl)-5-(trifluoromethyl)benzenesulfonamide
1g
97%
MFCD27665321
Videos
SDS