missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(4-methoxy-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64113.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15415665

  • £70.28 / 1g


Estimated Shipment date
from Supplier 10-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(4-methoxy-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
424.23
MFCD27975666

Specifications

3-Bromo-N-(4-methoxy-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
Solid
424.23
1g
97%
MFCD27975666
Videos
SDS