missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-[4-fluoro-3-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64720.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15465705

  • £42.72 / 1g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-[4-fluoro-3-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide
466.17
MFCD27665308

Specifications

3-Bromo-N-[4-fluoro-3-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide
97%
466.17
Solid
1g
MFCD27665308
Videos
SDS