missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(4-ethylphenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60693.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15457178

  • £358.49 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(4-ethylphenyl)benzenesulfonamide
C14H14BrNO2S
340.23
MFCD12567199

Specifications

3-Bromo-N-(4-ethylphenyl)benzenesulfonamide
C14H14BrNO2S
MFCD12567199
97%
340.23
1g
Videos
SDS