missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(4-chlorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64048.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458666

  • £249.10 / 5g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(4-chlorophenyl)-5-(trifluoromethyl)benzenesulfonamide
414.62
MFCD27665247

Specifications

3-Bromo-N-(4-chlorophenyl)-5-(trifluoromethyl)benzenesulfonamide
5g
414.62
Solid
97%
MFCD27665247
Videos
SDS