missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-[4-chloro-3-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64256.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15435675

  • £42.72 / 1g


Estimated Shipment date
from Supplier 09-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-[4-chloro-3-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide
482.62
MFCD27665267

Specifications

3-Bromo-N-[4-chloro-3-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide
1g
482.62
Solid
97%
MFCD27665267
Videos
SDS