missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(4-chloro-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64072.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15478686

  • £314.82 / 5g


Estimated Shipment date
from Supplier 11-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(4-chloro-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
428.65
MFCD27665251

Specifications

3-Bromo-N-(4-chloro-2-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
5g
428.65
Solid
97%
MFCD27665251
Videos
SDS