missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(4-chloro-2-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64669.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409066

  • £66.46 / 1g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(4-chloro-2-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
432.61
MFCD27665301

Specifications

3-Bromo-N-(4-chloro-2-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
1g
432.61
Solid
97%
MFCD27665301
Videos
SDS