missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(4-bromo-3-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64607.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15409016

  • £54.38 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(4-bromo-3-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
473.1
MFCD27665292

Specifications

3-Bromo-N-(4-bromo-3-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
1g
473.1
Solid
97%
MFCD27665292
Videos
SDS