missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(3-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64977.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419296

  • £85.44 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(3-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
394.21
MFCD27665333
3-Bromo-N-(m-tolyl)-5-(trifluoromethyl)benzenesulfonamide

Specifications

3-Bromo-N-(3-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
Solid
394.21
3-Bromo-N-(m-tolyl)-5-(trifluoromethyl)benzenesulfonamide
1g
97%
MFCD27665333
Videos
SDS