missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(3-methoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64712.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15419096

  • £54.38 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(3-methoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide
410.21
MFCD27975693

Specifications

3-Bromo-N-(3-methoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide
Solid
410.21
1g
97%
MFCD27975693
Videos
SDS