missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(3-fluorobenzyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64231.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15408796

  • £62.96 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(3-fluorobenzyl)-5-(trifluoromethyl)benzenesulfonamide
412.2
MFCD27975674

Specifications

3-Bromo-N-(3-fluorobenzyl)-5-(trifluoromethyl)benzenesulfonamide
97%
412.2
Solid
1g
MFCD27975674
Videos
SDS