missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-Bromo-N-(3-chloro-4-methoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%
Brand: Alfa Aesar™ H64323.06
Code : NEW
Additional Details : Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
3-Bromo-N-(3-chloro-4-methoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide | |
444.65 | |
MFCD27665271 |
Specifications
3-Bromo-N-(3-chloro-4-methoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide | |
5g | |
444.65 |
Solid | |
97% | |
MFCD27665271 |