missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(3-bromophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64896.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489236

  • £79.71 / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(3-bromophenyl)-5-(trifluoromethyl)benzenesulfonamide
459.08
MFCD27665322

Specifications

3-Bromo-N-(3-bromophenyl)-5-(trifluoromethyl)benzenesulfonamide
Solid
459.08
1g
97%
MFCD27665322
Videos
SDS