missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-Bromo-N-(3,5-dimethylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%
Brand: Alfa Aesar™ H64377.06
Code : NEW
Additional Details : Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
3-Bromo-N-(3,5-dimethylphenyl)-5-(trifluoromethyl)benzenesulfonamide | |
408.23 | |
MFCD27665275 |
Specifications
3-Bromo-N-(3,5-dimethylphenyl)-5-(trifluoromethyl)benzenesulfonamide | |
5g | |
408.23 |
Solid | |
97% | |
MFCD27665275 |