missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2-isopropylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64222.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15408786

  • £70.28 / 1g


Estimated Shipment date
from Supplier 30-01-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(2-isopropylphenyl)-5-(trifluoromethyl)benzenesulfonamide
422.26
MFCD27665266

Specifications

3-Bromo-N-(2-isopropylphenyl)-5-(trifluoromethyl)benzenesulfonamide
1g
422.26
Solid
97%
MFCD27665266
Videos
SDS