missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64930.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15489256

  • £331.78 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(2-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
398.17
MFCD27665326

Specifications

3-Bromo-N-(2-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
97%
398.17
Solid
5g
MFCD27665326
Videos
SDS