missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-[2-fluoro-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64496.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15495685

  • £298.92 / 5g


Estimated Shipment date
from Supplier 11-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-[2-fluoro-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide
466.17
MFCD27665285

Specifications

3-Bromo-N-[2-fluoro-5-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide
97%
466.17
Solid
5g
MFCD27665285
Videos
SDS