Alfa Aesar™ 3-Bromo-N-(2-fluoro-5-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H64117.MD

UNSPSC: 12352105

Code: NEW

Additional Details:
Additional Details: Weight: 0.00025kg


Quantity 250mg

Product Code. 15445665

Quantity Price
1 £ 14.95 / 250mg
Estimated Shipment date
from Supplier 13-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 3-Bromo-N-(2-fluoro-5-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
Formula Weight 412.2
MDL Number MFCD27665253

Specification

Formula Weight 412.2
MDL Number MFCD27665253
Quantity 250mg
Chemical Name or Material 3-Bromo-N-(2-fluoro-5-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
Assay 97%
Physical Form Solid
Documentation

Technical documents