missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2-ethylhexyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64565.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15478986

  • £99.64 / 1g


Estimated Shipment date
from Supplier 09-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(2-ethylhexyl)-5-(trifluoromethyl)benzenesulfonamide
416.3
MFCD27665290

Specifications

3-Bromo-N-(2-ethylhexyl)-5-(trifluoromethyl)benzenesulfonamide
1g
416.3
Solid
97%
MFCD27665290
Videos
SDS