missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2-ethoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64186.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15468756

  • £249.10 / 5g


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(2-ethoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide
424.23
MFCD27975671

Specifications

3-Bromo-N-(2-ethoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide
5g
424.23
Solid
97%
MFCD27975671
Videos
SDS