missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-[2-chloro-4-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64170.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15465665

  • £125.08 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-[2-chloro-4-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide
482.62
MFCD27975670

Specifications

3-Bromo-N-[2-chloro-4-(trifluoromethyl)phenyl]-5-(trifluoromethyl)benzenesulfonamide
1g
482.62
Solid
97%
MFCD27975670
Videos
SDS