missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2-bromo-4-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64068.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15448686

  • £298.92 / 5g


Estimated Shipment date
from Supplier 03-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(2-bromo-4-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
473.1
MFCD27665250

Specifications

3-Bromo-N-(2-bromo-4-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
5g
473.1
Solid
97%
MFCD27665250
Videos
SDS