missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2-bromo-4-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64015.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15458646

  • £259.70 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(2-bromo-4-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
477.07
MFCD27665243

Specifications

3-Bromo-N-(2-bromo-4-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
5g
477.07
Solid
97%
MFCD27665243
Videos
SDS