missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2,5-dimethoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64638.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15469046

  • £259.70 / 5g


Estimated Shipment date
from Supplier 06-06-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(2,5-dimethoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide
440.23
MFCD27665299

Specifications

3-Bromo-N-(2,5-dimethoxyphenyl)-5-(trifluoromethyl)benzenesulfonamide
5g
440.23
Solid
97%
MFCD27665299
Videos
SDS