missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2,5-difluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60885.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15436076

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(2,5-difluorophenyl)benzenesulfonamide
C12H8BrF2NO2S
348.16
MFCD08264781

Specifications

3-Bromo-N-(2,5-difluorophenyl)benzenesulfonamide
C12H8BrF2NO2S
MFCD08264781
97%
348.16
1g
Videos
SDS