missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2,4-dimethylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64131.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15488716

  • £62.96 / 1g


Estimated Shipment date
from Supplier 02-02-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(2,4-dimethylphenyl)-5-(trifluoromethyl)benzenesulfonamide
408.23
MFCD27665254

Specifications

3-Bromo-N-(2,4-dimethylphenyl)-5-(trifluoromethyl)benzenesulfonamide
1g
408.23
Solid
97%
MFCD27665254
Videos
SDS