missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2,4-difluorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64726.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15489106

  • £298.92 / 5g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(2,4-difluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
416.16
MFCD27665311

Specifications

3-Bromo-N-(2,4-difluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
5g
416.16
Solid
97%
MFCD27665311
Videos
SDS