Learn More
3-Bromo-7-nitroindazole, Tocris Bioscience™
Selective nNOS inhibitor
Brand:  Tocris (Bio-Techne) 073510
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487923
  • £128.25 / 10mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Potent inhibitor of rat cerebellar nitric oxide synthase; more potent than 7-nitroindazole (Cat. No. 0602).
Specifications
Chemical Identifiers
74209-34-0
242.032
1649
C7H4BrN3O2
NFSTZPMYAZRZPC-UHFFFAOYSA-N
Specifications
3-Bromo-7-nitroindazole
>98%
C7H4BrN3O2
C1=CC2=C(NN=C2C(=C1)[N+](=O)[O-])Br
242.032
242.03
74209-34-0
10mg
NFSTZPMYAZRZPC-UHFFFAOYSA-N
3-bromo-7-nitro-2H-indazole
1649
Videos