missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-methoxy-2-methylpyridine, 98%, Thermo Scientific™

Catalog Number p-7054075
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
126717-59-7
C7H8BrNO
202.051
MFCD03094943
VWNXCCTWVAQAPL-UHFFFAOYSA-N
5-bromo-2-methoxy-6-picoline, 3-bromo-6-methoxy-2-picoline, 3-bromo-6-methoxy-2-methyl-pyridine, 2-methoxy-5-bromo-6-picoline, 2-methyl-3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-6-methyl pyridine, 2-methoxy-5-bromo-6-methylpyridine, 5-bromo-2-methoxy-6-methylpyridine, 5-bromo-6-methyl-2-methoxypyridine, pyridine, 3-bromo-6-methoxy-2-methyl
10798146
3-bromo-6-methoxy-2-methylpyridine
CC1=C(C=CC(=N1)OC)Br
This item is not returnable. View return policy
126717-59-7
C7H8BrNO
202.051
MFCD03094943
VWNXCCTWVAQAPL-UHFFFAOYSA-N
5-bromo-2-methoxy-6-picoline, 3-bromo-6-methoxy-2-picoline, 3-bromo-6-methoxy-2-methyl-pyridine, 2-methoxy-5-bromo-6-picoline, 2-methyl-3-bromo-6-methoxypyridine, 5-bromo-2-methoxy-6-methyl pyridine, 2-methoxy-5-bromo-6-methylpyridine, 5-bromo-2-methoxy-6-methylpyridine, 5-bromo-6-methyl-2-methoxypyridine, pyridine, 3-bromo-6-methoxy-2-methyl
10798146
3-bromo-6-methoxy-2-methylpyridine
CC1=C(C=CC(=N1)OC)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 1 g
Formula Weight 202.05
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromo-6-methoxy-2-methylpyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'