missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-hydroxy-2-methylpyridine, 97%, Thermo Scientific™

Catalog Number 15495468
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated
TRUSTED_SUSTAINABILITY
CAS 54923-31-8
Melting Point 199°C to 203°C
Molecular Formula C6H6BrNO
MDL Number MFCD03427653
Quantity 1 g
Synonym 3-bromo-6-hydroxy-2-methylpyridine, 5-bromo-6-methylpyridin-2-ol, 3-bromo-6-hydroxy-2-picoline, 5-bromo-2-hydroxy-6-picoline, 5-bromo-6-methyl-2 1h-pyridinone, 2 1h-pyridinone, 5-bromo-6-methyl, 5-bromo-2-hydroxy-6-methylpyridine, 5-bromo-6-methyl-2-pyridinol, 5-bromo-6-methylpyridin-2 1h-one, acmc-20aiyp
Solubility Information Slightly soluble in water.
InChI Key UJHCRBDEJPQFIA-UHFFFAOYSA-N
SMILES CC1=C(Br)C=CC(=O)N1
IUPAC Name 5-bromo-6-methyl-1H-pyridin-2-one
Molecular Weight (g/mol) 188.02
PubChem CID 2734417
Formula Weight 188.02
Percent Purity 97%
Chemical Name or Material 3-Bromo-6-hydroxy-2-methylpyridine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'