missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-5-(trifluoromethyl)-N-[4-(trifluoromethyl)phenyl]benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64359.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15405685

  • £31.48 / 1g


Estimated Shipment date
from Supplier 08-05-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-5-(trifluoromethyl)-N-[4-(trifluoromethyl)phenyl]benzenesulfonamide
448.18
MFCD27665273

Specifications

3-Bromo-5-(trifluoromethyl)-N-[4-(trifluoromethyl)phenyl]benzenesulfonamide
97%
MFCD27665273
Solid
448.18
1g
Videos
SDS