missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3-Bromo-5-(trifluoromethyl)-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide, 97%
Brand: Alfa Aesar™ H64961.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
3-Bromo-5-(trifluoromethyl)-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide | |
448.18 | |
MFCD27665330 |
Specifications
3-Bromo-5-(trifluoromethyl)-N-[3-(trifluoromethyl)phenyl]benzenesulfonamide | |
97% | |
MFCD27665330 |
Solid | |
448.18 | |
1g |