missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-5-(trifluoromethyl)-N-(2,3,4-trifluorophenyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64577.03

Code : NEW

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15428996

  • £46.00 / 1g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-5-(trifluoromethyl)-N-(2,3,4-trifluorophenyl)benzenesulfonamide
434.15
MFCD27665291

Specifications

3-Bromo-5-(trifluoromethyl)-N-(2,3,4-trifluorophenyl)benzenesulfonamide
97%
MFCD27665291
Solid
434.15
1g
Videos
SDS