missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-5-(trifluoromethyl)benzoic acid, 97%

Brand:  Alfa Aesar™ H63776.06

Code : NEW

Additional Details : CAS Number : 328-67-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15470649

  • £14.80 / 5g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-5-(trifluoromethyl)benzoic acid
328-67-6
269.02
MFCD03412186

Specifications

3-Bromo-5-(trifluoromethyl)benzoic acid
Solid
269.02
5g
328-67-6
97%
MFCD03412186
Videos
SDS