missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-5-nitroanisole, 98%

Brand:  Alfa Aesar™ H64864.03

Code : NEW

Additional Details : CAS Number : 16618-67-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15462259

  • £23.96 / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-5-nitroanisole
232.03
1947332
16618-67-0
MFCD04112752
1-Bromo-3-methoxy-5-nitrobenzene

Specifications

3-Bromo-5-nitroanisole
85° to 88°C
Powder
98%
MFCD04112752
1-Bromo-3-methoxy-5-nitrobenzene
16618-67-0
Brown
1g
232.03
1947332
Videos
SDS