missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-methylaniline, 98%, Thermo Scientific Chemicals

Catalog Number p-7053821
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
74586-53-1
C7H8BrN
186.052
MFCD11845553
YIZRPAWCIFTHNA-UHFFFAOYSA-N
3-bromo-5-methylbenzenamine, 3-amino-5-bromotoluene, benzenamine, 3-bromo-5-methyl, 5-brom-3-toluidin, 5-bromo-m-toluidine, acmc-209ova, 3-bromo-5-methyl-aniline, 3-bromo-5-methyl aniline, ksc494e0l
3018526
3-bromo-5-methylaniline
CC1=CC(=CC(=C1)Br)N
This item is not returnable. View return policy
74586-53-1
C7H8BrN
186.052
MFCD11845553
YIZRPAWCIFTHNA-UHFFFAOYSA-N
3-bromo-5-methylbenzenamine, 3-amino-5-bromotoluene, benzenamine, 3-bromo-5-methyl, 5-brom-3-toluidin, 5-bromo-m-toluidine, acmc-209ova, 3-bromo-5-methyl-aniline, 3-bromo-5-methyl aniline, ksc494e0l
3018526
3-bromo-5-methylaniline
CC1=CC(=CC(=C1)Br)N

3-Bromo-5-methylaniline is used as pharmaceutical intermediates and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-5-methylaniline is used as pharmaceutical intermediates and chemical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color Brown
Quantity 1 g
UN Number UN2811
Solubility Information Slightly soluble in water.
Formula Weight 186.05
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Bromo-5-methylaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'