missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(methoxycarbonyl)benzeneboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number p-7034070
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1025718-78-8
C14H18BBrO4
341.008
MFCD12405334
YTLNMWYKZKGCOV-UHFFFAOYSA-N
methyl 3-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 3-bromo-5-methoxycarbonyl benzeneboronic acid pinacol ester, 3-bromo-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzoic acid methyl ester, 3-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzoic acid methyl ester, 3-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzoic acid, methyl ester, benzoic acid, 3-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, methyl ester
53436914
methyl 3-bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)Br)C(=O)OC
This item is not returnable. View return policy
1025718-78-8
C14H18BBrO4
341.008
MFCD12405334
YTLNMWYKZKGCOV-UHFFFAOYSA-N
methyl 3-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoate, 3-bromo-5-methoxycarbonyl benzeneboronic acid pinacol ester, 3-bromo-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzoic acid methyl ester, 3-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzoic acid methyl ester, 3-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzoic acid, methyl ester, benzoic acid, 3-bromo-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-, methyl ester
53436914
methyl 3-bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC(=C2)Br)C(=O)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 98°C to 102°C
Quantity 250 mg
Formula Weight 341.01
Percent Purity 97%
Chemical Name or Material 3-Bromo-5-(methoxycarbonyl)benzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'