missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(hydroxymethyl)pyridine, 97%, Thermo Scientific™

Catalog Number 11906461
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
37669-64-0
C6H6BrNO
188.02
MFCD03265757
WDVDHJLKXYCOFS-UHFFFAOYSA-N
5-bromopyridin-3-yl methanol, 5-bromo-3-pyridinemethanol, 3-bromo-5-hydroxymethylpyridine, 5-bromo-3-hydroxymethylpyridine, 5-bromo-3-pyridinyl methanol, 5-bromo-pyridin-3-yl-methanol, 3-pyridinemethanol, 5-bromo, 5-bromo-3-pyridyl methan-1-ol, 3-bromo-5-hydroxymethyl pyridine, 3-bromo-pyridin-5-yl methanol
2784733
(5-bromopyridin-3-yl)methanol
OCC1=CC(Br)=CN=C1
This item is not returnable. View return policy
37669-64-0
C6H6BrNO
188.02
MFCD03265757
WDVDHJLKXYCOFS-UHFFFAOYSA-N
5-bromopyridin-3-yl methanol, 5-bromo-3-pyridinemethanol, 3-bromo-5-hydroxymethylpyridine, 5-bromo-3-hydroxymethylpyridine, 5-bromo-3-pyridinyl methanol, 5-bromo-pyridin-3-yl-methanol, 3-pyridinemethanol, 5-bromo, 5-bromo-3-pyridyl methan-1-ol, 3-bromo-5-hydroxymethyl pyridine, 3-bromo-pyridin-5-yl methanol
2784733
(5-bromopyridin-3-yl)methanol
OCC1=CC(Br)=CN=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 27.0°C
Color Yellow
Boiling Point 154.0°C (4.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 188.02
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 3-Bromo-5-(hydroxymethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-5-(hydroxymethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'