missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorobenzotrifluoride, 97%, Thermo Scientific™

Catalog Number 10744672
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
130723-13-6
C7H3BrF4
243
MFCD00042498
LIGBGEJPUQBLTG-UHFFFAOYSA-N
3-bromo-5-fluorobenzotrifluoride, 1-bromo-3-fluoro-5-trifluoromethyl benzene, 3-bromo-5-fluorotrifluoromethylbenzene, 3-fluoro-5-trifluoromethyl bromobenzene, 5-fluoro-3-bromobenzotrifluoride, 3-fluoro-5-trifluoromethylbromobenzene, 3-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, 3-bromo-5-fluoro-trifluoromethylbenzene, benzene, 1-bromo-3-fluoro-5-trifluoromethyl
2736323
1-bromo-3-fluoro-5-(trifluoromethyl)benzene
C1=C(C=C(C=C1F)Br)C(F)(F)F
This item is not returnable. View return policy
130723-13-6
C7H3BrF4
243
MFCD00042498
LIGBGEJPUQBLTG-UHFFFAOYSA-N
3-bromo-5-fluorobenzotrifluoride, 1-bromo-3-fluoro-5-trifluoromethyl benzene, 3-bromo-5-fluorotrifluoromethylbenzene, 3-fluoro-5-trifluoromethyl bromobenzene, 5-fluoro-3-bromobenzotrifluoride, 3-fluoro-5-trifluoromethylbromobenzene, 3-bromo-alpha,alpha,alpha,5-tetrafluorotoluene, 3-bromo-5-fluoro-trifluoromethylbenzene, benzene, 1-bromo-3-fluoro-5-trifluoromethyl
2736323
1-bromo-3-fluoro-5-(trifluoromethyl)benzene
C1=C(C=C(C=C1F)Br)C(F)(F)F
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.5100g/mL
Boiling Point 146°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.51
Formula Weight 243
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 3-Bromo-5-fluorobenzotrifluoride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-5-fluorobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'