missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluoro-2-methoxypyridine, 95%, Thermo Scientific™

Catalog Number p-7056150
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
884494-81-9
C6H5BrFNO
206.014
MFCD04972397
HSWCOAGQYSBFAK-UHFFFAOYSA-N
3-bromo-5-fluoro-2-methoxypyrdine, 2-methoxy-3-bromo-5-fluoropyridine, synquest 4h07-b-01, 3-bromo-5-fluoro-2-methoxy-pyridine, pyridine,3-bromo-5-fluoro-2-methoxy, pyridine, 3-bromo-5-fluoro-2-methoxy, pubchem1175, acmc-209qt8, 3-bromo-5-fluoropyridin-2-yl methyl ether, 3-bromo-5-fluoro-2-methoxy pyridine
40427006
3-bromo-5-fluoro-2-methoxypyridine
COC1=C(C=C(C=N1)F)Br
This item is not returnable. View return policy
884494-81-9
C6H5BrFNO
206.014
MFCD04972397
HSWCOAGQYSBFAK-UHFFFAOYSA-N
3-bromo-5-fluoro-2-methoxypyrdine, 2-methoxy-3-bromo-5-fluoropyridine, synquest 4h07-b-01, 3-bromo-5-fluoro-2-methoxy-pyridine, pyridine,3-bromo-5-fluoro-2-methoxy, pyridine, 3-bromo-5-fluoro-2-methoxy, pubchem1175, acmc-209qt8, 3-bromo-5-fluoropyridin-2-yl methyl ether, 3-bromo-5-fluoro-2-methoxy pyridine
40427006
3-bromo-5-fluoro-2-methoxypyridine
COC1=C(C=C(C=N1)F)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Beilstein 15497859
Sensitivity Light sensitive
Formula Weight 206.01
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-fluoro-2-methoxypyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'