missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chloropyridine, 97%, ACROS Organics™

Brand:  Acros Organics 437400250

Code : 29

Additional Details : CAS Number : 73583-39-8 Weight : 0.07500kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III


Quantity 25g

 View more versions of this product

Product Code. 11916381

  • £387.80 / 25g


EU Stock 1
Add to basket


Specifications

Chemical Identifiers

3-Bromo-5-chloropyridine
96.0
97%
C5H3BrClN
73583-39-8
100.0
97%
192.44

Specifications

3-Bromo-5-chloropyridine
96.0
Authentic
97%
C5H3BrClN
192.44
78.7° to 80.1°C
73583-39-8
100.0
Crystalline Powder
97%
3% Max.
White to brown
25g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 3-Bromo-5-chloropyridine

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

SDS