missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-5-chlorophenol, ≥97%

Brand:  Alfa Aesar™ H64844.06

Code : NEW

Additional Details : CAS Number : 56962-04-0 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15412239

  • £45.70 / 5g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-5-chlorophenol
207.45
3234780
56962-04-0
MFCD07780676

Specifications

3-Bromo-5-chlorophenol
Solid
207.45
3234780
5g
56962-04-0
≥97%
MFCD07780676
66° to 70°C
Videos
SDS